Gene Transcripts

Gene Symbol
Rasl10b
Entrez Gene ID
303382
Gene Name
RAS-like, family 10, member B
Gene Aliases
Chromosome Location
Chr. 10: 70626210 - 70639072 on Build Rnor_6.0
UniGene ID
Rn.222772
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191648.1 NP_001178577.1 3 451
XM_006247028.2 XP_006247090.1 4 1013
XM_006247029.2 XP_006247091.1 5 1233
XM_006247030.2 XP_006247092.1 4 1230
XM_008768033.1 XP_008766255.1 4 2429
XM_008768034.1 XP_008766256.1 4 996

Gene Symbol
LOC103694863
Entrez Gene ID
103694863
Gene Name
ras-like protein family member 10B
Gene Aliases
Chromosome Location
Chr. Un|NW_007906801: 19835 - 32697 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773805.1 XP_008772027.1 4 2429
XM_008773806.1 XP_008772028.1 4 1013
XM_008773807.1 XP_008772029.1 5 1233
XM_008773808.1 XP_008772030.1 4 1230
XM_008773809.1 XP_008772031.1 3 917
XM_008773810.1 XP_008772032.1 4 996

View Full Product Details