Gene Transcripts

Gene Symbol
Msl1
Entrez Gene ID
303514
Gene Name
male-specific lethal 1 homolog (Drosophila)
Gene Aliases
RGD1564778
Chromosome Location
Chr. 10: 86710921 - 86722667 on Build Rnor_6.0
UniGene ID
Rn.22437
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107048.1 NP_001100518.1 3 1295
XM_006247304.2 XP_006247366.1 3 1614
GenBank mRNA FQ228290.1 - 3 1249

View Full Product Details