Gene Transcripts

Gene Symbol
Pnma2
Entrez Gene ID
305977
Gene Name
paraneoplastic Ma antigen 2
Gene Aliases
Chromosome Location
Chr. 15: 43581958 - 43589680 on Build Rnor_6.0
UniGene ID
Rn.150631
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107272.1 NP_001100742.1 3 1531
XM_006252116.2 XP_006252178.1 4 1705
XM_008770756.1 XP_008768978.1 3 1556

View Full Product Details