Gene Transcripts

Gene Symbol
Mak16
Entrez Gene ID
306526
Gene Name
MAK16 homolog (S. cerevisiae)
Gene Aliases
RGD1311297, Rbm13
Chromosome Location
Chr. 16: 64729254 - 64737807 on Build Rnor_6.0
UniGene ID
Rn.160904
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014002.1 NP_001014024.1 5 322
GenBank mRNA BC090035.1 - 5 322

View Full Product Details