Gene Transcripts

Gene Symbol
Ssc5d
Entrez Gene ID
308341
Gene Name
scavenger receptor cysteine rich family, 5 domains
Gene Aliases
RGD1565772
Chromosome Location
Chr. 1: 72443756 - 72462293 on Build Rnor_6.0
UniGene ID
Rn.68344
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134545.1 NP_001128017.1 10 2119
XM_006228257.1 XP_006228319.1 10 2865

View Full Product Details