Gene Transcripts

Gene Symbol
Ubqln3
Entrez Gene ID
308905
Gene Name
ubiquilin 3
Gene Aliases
Chromosome Location
Chr. 1: 169331643 - 169334093 on Build Rnor_6.0
UniGene ID
Rn.66649
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107543.2 NP_001101013.1 2 1569
GenBank mRNA BC161885.1 - 2 1569

View Full Product Details