Gene Transcripts

Gene Symbol
Med19
Entrez Gene ID
311165
Gene Name
mediator complex subunit 19
Gene Aliases
RGD1311926
Chromosome Location
Chr. 3: 72080630 - 72091150 on Build Rnor_6.0
UniGene ID
Rn.233785
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107741.1 NP_001101211.1 2 325

View Full Product Details