Gene Transcripts
- Gene Symbol
- Ano3
- Entrez Gene ID
- 311287
- Gene Name
- anoctamin 3
- Gene Aliases
- Tmem16c
- Chromosome Location
- Chr. 3: 101843516 - 102151493 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_230381.8 |
XP_230381.7
|
10 |
1179 |
|
View Full Product Details