Gene Transcripts
- Gene Symbol
- PPFIA2
- Entrez Gene ID
- 8499
- Gene Name
- PTPRF interacting protein alpha 2
- Gene Aliases
- Chromosome Location
- Chr. 12: 81257975 - 81759748 on Build GRCh38
- UniGene ID
- Hs.506216
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001220473.2 |
NP_001207402.1
|
2, 3 |
553 |
|
NM_001220475.2 |
NP_001207404.1
|
2, 3 |
365 |
|
NM_001220476.2 |
NP_001207405.1
|
2, 3 |
365 |
|
NM_003625.4 |
NP_003616.2
|
3, 4 |
416 |
|
XM_017020079.1 |
XP_016875568.1
|
3, 4 |
400 |
|
XM_017020080.1 |
XP_016875569.1
|
2, 3 |
547 |
|
XM_017020081.1 |
XP_016875570.1
|
2, 3 |
547 |
|
XM_017020082.1 |
XP_016875571.1
|
3, 4 |
400 |
|
XM_017020083.1 |
XP_016875572.1
|
2, 3 |
547 |
|
XM_017020084.1 |
XP_016875573.1
|
2, 3 |
547 |
|
XM_017020085.1 |
XP_016875574.1
|
2, 3 |
547 |
|
XM_017020086.1 |
XP_016875575.1
|
3, 4 |
400 |
|
XM_017020087.1 |
XP_016875576.1
|
2, 3 |
534 |
|
XM_017020090.1 |
XP_016875579.1
|
3, 4 |
400 |
|
XM_017020091.1 |
XP_016875580.1
|
2, 3 |
547 |
|
XM_017020092.1 |
XP_016875581.1
|
2, 3 |
547 |
|
XM_017020093.1 |
XP_016875582.1
|
2, 3 |
547 |
|
XM_017020094.1 |
XP_016875583.1
|
3, 4 |
400 |
|
XM_017020095.1 |
XP_016875584.1
|
2, 3 |
547 |
|
XM_017020096.1 |
XP_016875585.1
|
2, 3 |
547 |
|
XM_017020097.1 |
XP_016875586.1
|
3, 4 |
400 |
|
XM_017020098.1 |
XP_016875587.1
|
3, 4 |
400 |
|
XM_017020100.1 |
XP_016875589.1
|
2, 3 |
547 |
|
XM_017020101.1 |
XP_016875590.1
|
3, 4 |
400 |
|
XM_017020102.1 |
XP_016875591.1
|
2, 3 |
534 |
|
XM_017020103.1 |
XP_016875592.1
|
2, 3 |
547 |
|
XM_017020104.1 |
XP_016875593.1
|
2, 3 |
547 |
|
XM_017020105.1 |
XP_016875594.1
|
2, 3 |
547 |
|
XM_017020106.1 |
XP_016875595.1
|
2, 3 |
547 |
|
XM_017020107.1 |
XP_016875596.1
|
3, 4 |
400 |
|
XM_017020108.1 |
XP_016875597.1
|
3, 4 |
400 |
|
XM_017020109.1 |
XP_016875598.1
|
2, 3 |
547 |
|
XM_017020111.1 |
XP_016875600.1
|
2, 3 |
547 |
|
GenBank mRNA |
AB210009.1 |
- |
2, 3 |
551 |
|
AF034799.1 |
- |
3, 4 |
424 |
|
BC104912.1 |
- |
2, 3 |
349 |
8143915 |
BC143485.1 |
- |
2, 3 |
349 |
9051993 |
DA499362.1 |
- |
2, 3 |
554 |
|
View Full Product Details