Gene Transcripts

Gene Symbol
Mettl7a
Entrez Gene ID
315306
Gene Name
methyltransferase like 7A
Gene Aliases
RGD1308407
Chromosome Location
Chr. 7: 141973553 - 141981432 on Build Rnor_6.0
UniGene ID
Rn.3341
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037355.1 NP_001032432.1 2 642
GenBank mRNA BC104688.1 - 1 378
BC105760.1 - 2 642 5623503

View Full Product Details