Gene Transcripts

Gene Symbol
Magee1
Entrez Gene ID
317232
Gene Name
melanoma antigen, family E, 1
Gene Aliases
DAMAGE, RGD1560259
Chromosome Location
Chr. X: 76083553 - 76087100 on Build Rnor_6.0
UniGene ID
Rn.38053
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079891.1 NP_001073360.1 1 1676
GenBank mRNA BC128754.1 - 1 1676

View Full Product Details