Gene Transcripts

Gene Symbol
Etv4
Entrez Gene ID
360635
Gene Name
ets variant 4
Gene Aliases
Pea3
Chromosome Location
Chr. 10: 89685058 - 89700314 on Build Rnor_6.0
UniGene ID
Rn.195602
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768086.1 XP_008766308.1 6 716

View Full Product Details