Gene Transcripts
- Gene Symbol
- Lmo7
- Entrez Gene ID
- 361084
- Gene Name
- LIM domain 7
- Gene Aliases
- Chromosome Location
- Chr. 15: 86242911 - 86458512 on Build Rnor_6.0
- UniGene ID
- Rn.98417
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001515.1 |
NP_001001515.1
|
3, 4 |
205 |
|
XM_006252397.2 |
XP_006252459.1
|
3, 4 |
441 |
|
XM_006252399.2 |
XP_006252461.1
|
3, 4 |
437 |
|
XM_006252400.2 |
XP_006252462.1
|
3, 4 |
437 |
|
XM_006252401.2 |
XP_006252463.1
|
3, 4 |
437 |
|
XM_008770874.1 |
XP_008769096.1
|
4, 5 |
337 |
|
XM_008770875.1 |
XP_008769097.1
|
3, 4 |
442 |
|
XM_008770876.1 |
XP_008769098.1
|
3, 4 |
442 |
|
XM_008770877.1 |
XP_008769099.1
|
3, 4 |
441 |
|
XM_008770879.1 |
XP_008769101.1
|
3, 4 |
441 |
|
GenBank mRNA |
AY533473.1 |
- |
3, 4 |
205 |
|
AY533474.1 |
- |
3, 4 |
205 |
|
View Full Product Details