Gene Transcripts

Gene Symbol
MTMR1
Entrez Gene ID
8776
Gene Name
myotubularin related protein 1
Gene Aliases
Chromosome Location
Chr. X: 150692997 - 150765103 on Build GRCh38
UniGene ID
Hs.347187
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306144.1 NP_001293073.1 2 355
NM_001306145.1 NP_001293074.1 2 355
NM_003828.3 NP_003819.1 2 355
XM_005274765.3 XP_005274822.1 2 317
XM_005274766.3 XP_005274823.1 2 316
XM_005274767.3 XP_005274824.1 2 335
XM_006724855.3 XP_006724918.1 2 355
XM_011531209.2 XP_011529511.1 2 115
XM_011531210.2 XP_011529512.1 2 319
XM_017029920.1 XP_016885409.1 2 317
XM_017029921.1 XP_016885410.1 1 30
XM_017029922.1 XP_016885411.1 2 111
XM_017029923.1 XP_016885412.1 1 18
XM_017029924.1 XP_016885413.1 2 335
GenBank mRNA AF057354.1 - 2 179
AJ224979.1 - 2 181
AK226109.1 - 2 309
AK295738.1 - 2 150
BC032947.1 - 2 352 4837744
DR000536.1 - 2 357

View Full Product Details