Gene Transcripts

Gene Symbol
Ndufb7
Entrez Gene ID
361385
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7
Gene Aliases
Chromosome Location
Chr. 19: 24701067 - 24705405 on Build Rnor_6.0
UniGene ID
Rn.105421
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108442.1 NP_001101912.1 2 277
GenBank mRNA FQ217491.1 - 2 237
FQ223670.1 - 2 239

View Full Product Details