Gene Transcripts
- Gene Symbol
- RGS11
- Entrez Gene ID
- 8786
- Gene Name
- regulator of G-protein signaling 11
- Gene Aliases
- RS11
- Chromosome Location
- Chr. 16: 268301 - 275943 on Build GRCh38
- UniGene ID
- Hs.65756
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286485.1 |
NP_001273414.1
|
5, 6 |
436 |
|
NM_001286486.1 |
NP_001273415.1
|
5, 6 |
373 |
|
NM_003834.2 |
NP_003825.1
|
6, 7 |
480 |
|
NM_183337.2 |
NP_899180.1
|
6, 7 |
469 |
|
XM_011522719.2 |
XP_011521021.1
|
7, 8 |
563 |
|
XM_011522720.2 |
XP_011521022.1
|
7, 8 |
503 |
|
XM_011522721.1 |
XP_011521023.1
|
7, 8 |
563 |
|
XM_011522722.1 |
XP_011521024.1
|
7, 8 |
563 |
|
XM_011522723.1 |
XP_011521025.1
|
6, 7 |
467 |
|
GenBank mRNA |
AB016929.1 |
- |
5, 6 |
404 |
|
AB209582.1 |
- |
1, 2 |
922 |
|
AF035153.1 |
- |
6, 7 |
440 |
|
AF035154.1 |
- |
6, 7 |
451 |
|
AK091701.1 |
- |
5, 6 |
370 |
|
AK293564.1 |
- |
6, 7 |
463 |
|
AK294448.1 |
- |
6, 7 |
463 |
|
AK308972.1 |
- |
5, 6 |
690 |
|
BC040504.1 |
- |
5, 6 |
1217 |
|
BC141803.1 |
- |
6, 7 |
444 |
40147243 |
View Full Product Details