Gene Transcripts

Gene Symbol
Prodh2
Entrez Gene ID
361538
Gene Name
proline dehydrogenase (oxidase) 2
Gene Aliases
Chromosome Location
Chr. 1: 88955125 - 88968501 on Build Rnor_6.0
UniGene ID
Rn.4247
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038588.1 NP_001033677.1 6 818
GenBank mRNA AF222852.1 - 6 808
BC158806.1 - 6 818 7324442

View Full Product Details