Gene Transcripts

Gene Symbol
Nudt8
Entrez Gene ID
361692
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 8
Gene Aliases
Chromosome Location
Chr. 1: 219247551 - 219249154 on Build Rnor_6.0
UniGene ID
Rn.23628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760189.1 XP_008758411.1 2 448
XM_341975.7 XP_341976.4 2 448

View Full Product Details