Gene Transcripts

Gene Symbol
Ndufs5
Entrez Gene ID
362588
Gene Name
NADH dehydrogenase (ubiquinone) Fe-S protein 5
Gene Aliases
Ndufs5b
Chromosome Location
Chr. 5: 141390577 - 141405504 on Build Rnor_6.0
UniGene ID
Rn.156413
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030052.1 NP_001025223.1 3 323
XM_006238827.2 XP_006238889.1 3 336
XM_006238828.2 XP_006238890.1 3 297
GenBank mRNA BC168721.1 - 3 301
FQ224355.1 - 3 281

Gene Symbol
LOC100363268
Entrez Gene ID
100363268
Gene Name
rCG31129-like
Gene Aliases
Chromosome Location
Chr. 2: 195940666 - 195941140 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729080.3 XP_002729126.1 1 278

View Full Product Details