Gene Transcripts

Gene Symbol
RGD1304554
Entrez Gene ID
363043
Gene Name
LOC363043
Gene Aliases
Pate-E
Chromosome Location
Chr. 8: 37014747 - 37016116 on Build Rnor_6.0
UniGene ID
Rn.105521
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108758.1 NP_001102228.1 3 279
GenBank mRNA JF412805.1 - 3 251

View Full Product Details