Gene Transcripts

Gene Symbol
Olr87
Entrez Gene ID
365329
Gene Name
olfactory receptor 87
Gene Aliases
Chromosome Location
Chr. 1: 168390615 - 168391553 on Build Rnor_6.0
UniGene ID
Rn.141899
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000543.1 NP_001000543.1 1 647

View Full Product Details