Gene Transcripts

Gene Symbol
Lhx8
Entrez Gene ID
365963
Gene Name
LIM homeobox 8
Gene Aliases
Lhx7
Chromosome Location
Chr. 2: 260572827 - 260598744 on Build Rnor_6.0
UniGene ID
Rn.133072
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012219.1 NP_001012219.1 3, 4 427
XM_006233528.2 XP_006233590.1 4, 5 693
GenBank mRNA BC090011.1 - 3, 4 427

View Full Product Details