Gene Transcripts

Gene Symbol
Med9
Entrez Gene ID
497914
Gene Name
mediator complex subunit 9
Gene Aliases
RGD1563669
Chromosome Location
Chr. 10: 46314639 - 46329374 on Build Rnor_6.0
UniGene ID
Rn.1743
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127302.1 NP_001120774.1 1 191
GenBank mRNA BC166759.1 - 1 191

View Full Product Details