Gene Transcripts
- Gene Symbol
- Lgr6
- Entrez Gene ID
- 498233
- Gene Name
- leucine-rich repeat-containing G protein-coupled receptor 6
- Gene Aliases
- RGD1560339
- Chromosome Location
- Chr. 13: 51811904 - 51931194 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_573455.6 |
XP_573455.1
|
6, 7 |
942 |
|
View Full Product Details