Gene Transcripts

Gene Symbol
RatNP-3b
Entrez Gene ID
498659
Gene Name
defensin RatNP-3 precursor
Gene Aliases
NP-3, NP-3a, NP-3b, Np3
Chromosome Location
Chr. 16: 75528348 - 75530511 on Build Rnor_6.0
UniGene ID
Rn.114810
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079898.2 NP_001073367.2 3 273
GenBank mRNA U16683.1 - 3 262

View Full Product Details