Gene Transcripts

Gene Symbol
Mfap1a
Entrez Gene ID
499878
Gene Name
microfibrillar-associated protein 1A
Gene Aliases
RGD1562232
Chromosome Location
Chr. 3: 113427229 - 113445458 on Build Rnor_6.0
UniGene ID
Rn.93042
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191964.1 NP_001178893.1 5 984

View Full Product Details