Gene Transcripts
- Gene Symbol
- VPS4B
- Entrez Gene ID
- 9525
- Gene Name
- vacuolar protein sorting 4 homolog B
- Gene Aliases
- MIG1, SKD1, SKD1B, VPS4-2
- Chromosome Location
- Chr. 18: 63389190 - 63422519 on Build GRCh38
- UniGene ID
- Hs.126550
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004869.3 |
NP_004860.2
|
8, 9 |
1135 |
|
XM_006722582.2 |
XP_006722645.1
|
7, 8 |
1011 |
|
XM_011526268.2 |
XP_011524570.1
|
8, 9 |
1135 |
|
GenBank mRNA |
AF038960.1 |
- |
8, 9 |
1064 |
|
AF195514.1 |
- |
8, 9 |
1076 |
|
AK290932.1 |
- |
8, 9 |
1079 |
|
AK291253.1 |
- |
8, 9 |
1033 |
|
AK308892.1 |
- |
8, 9 |
1099 |
|
AK309322.1 |
- |
7, 8 |
1011 |
|
AK311041.1 |
- |
7, 8 |
1011 |
|
AY232629.1 |
- |
8, 9 |
885 |
|
BC039574.1 |
- |
8, 9 |
1042 |
6042862 |
View Full Product Details