Gene Transcripts

Gene Symbol
LOC679924
Entrez Gene ID
679924
Gene Name
similar to Kif19A CG9913-PB, isoform B
Gene Aliases
Chromosome Location
Chr. 12: 16233512 - 16283177 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006248973.2 XP_006249035.1 12 1680

View Full Product Details