Gene Transcripts

Gene Symbol
C14orf2
Entrez Gene ID
9556
Gene Name
chromosome 14 open reading frame 2
Gene Aliases
MP68, PLPM
Chromosome Location
Chr. 14: 103912288 - 103921566 on Build GRCh38
UniGene ID
Hs.109052
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127393.1 NP_001120865.1 3 424
NM_004894.2 NP_004885.1 2 155
GenBank mRNA AF054175.1 - 2 118
AF116639.1 - 151
AK309355.1 - 2 116
AK309499.1 - 2 118
AK310120.1 - 2 109
AK312099.1 - 2 116
BC000429.2 - 2 107 2819801
BC001944.1 - 2 107 2819801
BX161388.1 - 3 359
CB127906.1 - 3 383
DA526965.1 - 2 155

View Full Product Details