Gene Transcripts

Gene Symbol
Nme4
Entrez Gene ID
685679
Gene Name
non-metastatic cells 4, protein expressed in
Gene Aliases
Chromosome Location
Chr. 10: 15461567 - 15465422 on Build Rnor_6.0
UniGene ID
Rn.145946
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109478.1 NP_001102948.1 5 510
GenBank mRNA CB724692.1 - 4 393

View Full Product Details