Gene Transcripts

Gene Symbol
Nudt11
Entrez Gene ID
680248
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 11
Gene Aliases
Chromosome Location
Chr. X: 17171510 - 17215108 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006256765.1 XP_006256827.1 2 1261

View Full Product Details