Gene Transcripts

Gene Symbol
Nipal3
Entrez Gene ID
502990
Gene Name
NIPA-like domain containing 3
Gene Aliases
Npal3, RGD1563439
Chromosome Location
Chr. 5: 153808181 - 153843951 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239277.2 XP_006239339.1 7 955
XM_578498.5 XP_578498.3 7 1022

View Full Product Details