Gene Transcripts
- Gene Symbol
- LAPTM4A
- Entrez Gene ID
- 9741
- Gene Name
- lysosomal protein transmembrane 4 alpha
- Gene Aliases
- HUMORF13, LAPTM4, MBNT, Mtrp
- Chromosome Location
- Chr. 2: 20032650 - 20052028 on Build GRCh38
- UniGene ID
- Hs.467807
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014713.4 |
NP_055528.1
|
1, 2 |
617 |
|
GenBank mRNA |
AK129875.1 |
- |
1, 2 |
267 |
|
AK304433.1 |
- |
1, 2 |
215 |
|
AY359028.1 |
- |
1, 2 |
233 |
|
BC000421.2 |
- |
1, 2 |
194 |
2964450 |
BC003158.1 |
- |
1, 2 |
257 |
2958870 |
BC093075.1 |
- |
1, 2 |
217 |
30402963 |
CB962971.1 |
- |
|
279 |
|
CR456758.1 |
- |
1, 2 |
109 |
|
D14696.1 |
- |
1, 2 |
255 |
|
DR156048.1 |
- |
1, 2 |
650 |
|
View Full Product Details