Gene Transcripts

Gene Symbol
Vom2r54
Entrez Gene ID
691438
Gene Name
vomeronasal 2 receptor, 54
Gene Aliases
Chromosome Location
Chr. 7: 16931774 - 16966830 on Build Rnor_6.0
UniGene ID
Rn.220928
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099494.1 NP_001092964.1 3 1266
XM_006241129.1 XP_006241191.1 4 1485

View Full Product Details