Gene Transcripts

Gene Symbol
Nt5c
Entrez Gene ID
100360582
Gene Name
5', 3'-nucleotidase, cytosolic
Gene Aliases
Umph2
Chromosome Location
Chr. 10: 104052807 - 104054805 on Build Rnor_6.0
UniGene ID
Rn.199091
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271189.1 NP_001258118.1 4 412
GenBank mRNA FQ227491.1 - 4 423
FQ233678.1 - 4 412

View Full Product Details