Gene Transcripts
- Gene Symbol
- NPM2
- Entrez Gene ID
- 10361
- Gene Name
- nucleophosmin/nucleoplasmin 2
- Gene Aliases
- Chromosome Location
- Chr. 8: 22023308 - 22036908 on Build GRCh38
- UniGene ID
- Hs.131055
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286680.1 |
NP_001273609.1
|
4, 5 |
791 |
|
NM_001286681.1 |
NP_001273610.1
|
3, 4 |
377 |
|
NM_182795.1 |
NP_877724.1
|
3, 4 |
377 |
|
XM_011544360.2 |
XP_011542662.1
|
2, 3 |
1013 |
|
XM_011544362.2 |
XP_011542664.1
|
4, 5 |
410 |
|
XM_011544363.2 |
XP_011542665.1
|
3, 4 |
291 |
|
XM_011544364.2 |
XP_011542666.1
|
3, 4 |
700 |
|
XM_017012948.1 |
XP_016868437.1
|
4, 5 |
1010 |
|
XM_017012949.1 |
XP_016868438.1
|
3, 4 |
1176 |
|
GenBank mRNA |
AK094267.1 |
- |
3, 4 |
1152 |
|
AY262113.1 |
- |
4, 5 |
467 |
|
AY262114.1 |
- |
3, 4 |
377 |
|
BC068078.1 |
- |
3, 4 |
369 |
6087597 |
View Full Product Details