Gene Transcripts

Gene Symbol
Lrit2
Entrez Gene ID
239038
Gene Name
leucine-rich repeat, immunoglobulin-like and transmembrane domains 2
Gene Aliases
A930010E21Rik, Lrrc22
Chromosome Location
Chr. 14: 37068049 - 37073735 on Build GRCm38
UniGene ID
Mm.134302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173418.3 NP_775594.1 2 197
GenBank mRNA AK044383.1 - 2 198
AK044611.1 - 2 199
BC037728.1 - 2 97
BC057628.1 - 2 172 4511498

View Full Product Details