Gene Transcripts

Gene Symbol
Spin2c
Entrez Gene ID
278240
Gene Name
spindlin family, member 2C
Gene Aliases
BC068162, SPIN-2C, Spin2
Chromosome Location
Chr. X: 153832002 - 153834243 on Build GRCm38
UniGene ID
Mm.293363
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005370.1 NP_001005370.1 2 896
GenBank mRNA BC068162.1 - 2 896

View Full Product Details