Gene Transcripts

Gene Symbol
KLRA1P
Entrez Gene ID
10748
Gene Name
killer cell lectin like receptor A1, pseudogene
Gene Aliases
KLRA1, KLRAP1, LY49L, Ly-49L, Ly49
Chromosome Location
Chr. 12: 10588478 - 10599835 on Build GRCh38
UniGene ID
Hs.159297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028045.1 4 606
GenBank mRNA AF047445.1 - 4 336
AK290077.1 - 3 307
AK296103.1 - 4 343
AK304677.1 - 3 302
AY334570.1 - 2 150
BC069672.1 - 4 309 7262588
BC069697.1 - 4 309 7262600
BC069734.1 - 4 309 7262612
BC101471.1 - 4 323 8068977
BC101473.1 - 4 326 8068979

View Full Product Details