Gene Transcripts

Gene Symbol
Olr35
Entrez Gene ID
293140
Gene Name
olfactory receptor 35
Gene Aliases
Chromosome Location
Chr. 1: 164706276 - 164707195 on Build Rnor_6.0
UniGene ID
Rn.103618
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000121.1 NP_001000121.1 1 338
GenBank mRNA BC086526.1 - 352

View Full Product Details