Gene Transcripts

Gene Symbol
Metrnl
Entrez Gene ID
316842
Gene Name
meteorin, glial cell differentiation regulator-like
Gene Aliases
Chromosome Location
Chr. 10: 110893261 - 110907256 on Build Rnor_6.0
UniGene ID
Rn.64557
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014104.1 NP_001014126.1 2 575
GenBank mRNA AB646250.1 - 3 328
BC086590.1 - 2 575 7310123

View Full Product Details