Gene Transcripts

Gene Symbol
PROSC
Entrez Gene ID
11212
Gene Name
proline synthetase cotranscribed homolog (bacterial)
Gene Aliases
Chromosome Location
Chr. 8: 37761753 - 37779768 on Build GRCh38
UniGene ID
Hs.304792
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007198.3 NP_009129.1 5 498
GenBank mRNA AK021923.1 - 5 366
AK291135.1 - 5 436
AL136616.1 - 5 478
BC012334.1 - 5 488 3546307
CR533532.1 - 5 421
DA086552.1 - 5 498

View Full Product Details