Gene Transcripts

Gene Symbol
Nipbl
Entrez Gene ID
294787
Gene Name
Nipped-B homolog (Drosophila)
Gene Aliases
IDN3
Chromosome Location
Chr. 2: 57508519 - 57676197 on Build Rnor_6.0
UniGene ID
Rn.225928
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760809.1 XP_008759031.1 26 5581

View Full Product Details