Gene Transcripts
- Gene Symbol
- LOC107984138
- Entrez Gene ID
- 107984138
- Gene Name
- serine/threonine-protein kinase SMG1-like
- Gene Aliases
- Chromosome Location
- Chr. 16: 70226339 - 70246610 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- SMG1
- Entrez Gene ID
- 23049
- Gene Name
- SMG1, nonsense mediated mRNA decay associated PI3K related kinase
- Gene Aliases
- 61E3.4, ATX, LIP
- Chromosome Location
- Chr. 16: 18804853 - 18926417 on Build GRCh38
- UniGene ID
- Hs.460179
- Species
- Homo sapiens
- Gene Symbol
- SMG1P1
- Entrez Gene ID
- 641298
- Gene Name
- SMG1P1, nonsense mediated mRNA decay associated PI3K related kinase pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 16: 22437008 - 22492220 on Build GRCh38
- UniGene ID
- Hs.552700
- Species
- Homo sapiens
- Gene Symbol
- SMG1P2
- Entrez Gene ID
- 440354
- Gene Name
- SMG1P2, nonsense mediated mRNA decay associated PI3K related kinase pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 16: 29545011 - 29613717 on Build GRCh38
- UniGene ID
- Hs.552700
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096982.1 |
- |
6 |
875 |
|
- Gene Symbol
- SMG1P3
- Entrez Gene ID
- 100271836
- Gene Name
- SMG1P3, nonsense mediated mRNA decay associated PI3K related kinase pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 16: 21446683 - 21502281 on Build GRCh38
- UniGene ID
- Hs.552700
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027155.2 |
|
22 |
3530 |
|
- Gene Symbol
- SMG1P6
- Entrez Gene ID
- 100422558
- Gene Name
- SMG1P6, nonsense mediated mRNA decay associated PI3K related kinasepseudogene 6
- Gene Aliases
- Chromosome Location
- Chr. 16: 29425411 - 29447026 on Build GRCh38
- UniGene ID
- Hs.552700
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135312.1 |
|
12 |
1616 |
|
- Gene Symbol
- SMG1P7
- Entrez Gene ID
- 100506060
- Gene Name
- SMG1P7, nonsense mediated mRNA decay associated PI3K related kinase pseudogene 7
- Gene Aliases
- Chromosome Location
- Chr. 16: 70219581 - 70226033 on Build GRCh38
- UniGene ID
- Hs.655258
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK308867.1 |
- |
9 |
1034 |
|
View Full Product Details