Gene Transcripts

Gene Symbol
NRBP2
Entrez Gene ID
340371
Gene Name
nuclear receptor binding protein 2
Gene Aliases
TRG16, pp9320
Chromosome Location
Chr. 8: 143829776 - 143840974 on Build GRCh38
UniGene ID
Hs.521926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178564.3 NP_848659.2 15 1424
XM_017013378.1 XP_016868867.1 15 1424
XM_017013379.1 XP_016868868.1 15 1424
XM_017013380.1 XP_016868869.1 15 1424
XM_017013381.1 XP_016868870.1 15 1424
XM_017013382.1 XP_016868871.1 15 1424
XM_017013383.1 XP_016868872.1 15 1424
GenBank mRNA AF318376.1 - 5 764
AK094017.1 - 2 133
AK293943.1 - 15 1424
AK308967.1 - 2 115
AL137662.1 - 13 1353
AL834530.1 - 8 663
AY277601.1 - 5 751
BC071605.1 - 14 1587
BC113873.1 - 12 1428 40053695
BC114481.1 - 13 1325
CR457350.1 - 7 555

View Full Product Details