Gene Transcripts

Gene Symbol
NDUFB2
Entrez Gene ID
4708
Gene Name
NADH:ubiquinone oxidoreductase subunit B2
Gene Aliases
AGGG, CI-AGGG
Chromosome Location
Chr. 7: 140696681 - 140706646 on Build GRCh38
UniGene ID
Hs.655788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004546.2 NP_004537.1 4 459
GenBank mRNA AF050639.1 - 4 452
AF067166.1 - 4 459
BC001168.1 - 4 426
BC063026.1 - 5 624 6064434

View Full Product Details