Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052971.2 | NP_443203.1 | 3 | 437 | |
GenBank mRNA | AF143867.1 | - | 2 | 869 | |
AJ306405.1 | - | 3 | 437 | ||
AJ409065.1 | - | 3 | 1781 | ||
AY052782.1 | - | 3 | 383 | ||
BC070199.1 | - | 3 | 435 | 4711742 | |
BX443829.2 | - | 3 | 425 |