Gene Transcripts

Gene Symbol
OCSTAMP
Entrez Gene ID
128506
Gene Name
osteoclast stimulatory transmembrane protein
Gene Aliases
C20orf123, OC-STAMP, dJ257E24.3
Chromosome Location
Chr. 20: 46541031 - 46550574 on Build GRCh38
UniGene ID
Hs.526636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080721.2 NP_542452.1 2 1050
XM_011528532.2 XP_011526834.1 1 860

View Full Product Details