Gene Transcripts
- Gene Symbol
- RPH3A
- Entrez Gene ID
- 22895
- Gene Name
- rabphilin 3A
- Gene Aliases
- Chromosome Location
- Chr. 12: 112791538 - 112898881 on Build GRCh38
- UniGene ID
- Hs.21239
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143854.1 |
NP_001137326.1
|
20, 21 |
2261 |
|
NM_014954.3 |
NP_055769.2
|
19, 20 |
2249 |
|
XM_011538048.1 |
XP_011536350.1
|
20, 21 |
2065 |
|
XM_017019034.1 |
XP_016874523.1
|
12, 13 |
1519 |
|
GenBank mRNA |
AB023202.1 |
- |
20, 21 |
2182 |
|
AK295696.1 |
- |
20, 21 |
2215 |
|
AK296296.1 |
- |
18, 19 |
1800 |
|
AK316112.1 |
- |
20, 21 |
2096 |
|
BC017259.2 |
- |
19, 20 |
2056 |
3510158 |
BX647281.1 |
- |
12, 13 |
2855 |
|
View Full Product Details