Gene Transcripts

Gene Symbol
PRICKLE4
Entrez Gene ID
29964
Gene Name
prickle planar cell polarity protein 4
Gene Aliases
C6orf49, OBTP, OEBT, TOMM6
Chromosome Location
Chr. 6: 41780762 - 41787372 on Build GRCh38
UniGene ID
Hs.715653
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013397.5 NP_037529.3 5 598
GenBank mRNA BC110458.1 - 3 1050
BC110459.1 - 3 1050 40024767
DA917216.1 - 1 212

View Full Product Details